CAS RN 968-21-8
IUPAC Navn:
(2S)-2-[(2S)-2-amino-4-methylpentanamido]-3-(4-hydroxyphenyl)propanoic acid
Synonymer:
(2S)-2-[(2S)-2-amino-4-methylpentanamido]-3-(4-hydroxyphenyl)propanoic acid
Molekylvægt (g/mol):
294.35
Molekylformel:
C15H22N2O4
InChi Key:
LHSGPCFBGJHPCY-STQMWFEESA-N
SMILES:
CC(C)C[C@H](N)C(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
1
–
3
af
3
Resultater
1
L-Leucyl-L-tyrosine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
L-Leucyl-L-tyrosine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
L-Leucyl-L-tyrosine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.