CAS RN 93882-12-3
IUPAC Navn:
sodium 6-{6-amino-8-[(4-chlorophenyl)sulfanyl]-9H-purin-9-yl}-7-hydroxy-2-oxo-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate
Synonymer:
sodium 6-{6-amino-8-[(4-chlorophenyl)sulfanyl]purin-9-yl}-7-hydroxy-2-oxo-tetrahydro-4H-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate
Molekylvægt (g/mol):
493.79
Molekylformel:
C16H14ClN5NaO6PS
InChi Key:
YIJFVHMIFGLKQL-UHFFFAOYNA-M
SMILES:
[Na+].NC1=C2N=C(SC3=CC=C(Cl)C=C3)N(C3OC4COP([O-])(=O)OC4C3O)C2=NC=N1
1
–
3
af
3
Resultater
1
8-CPT-cAMP sodium salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
8-CPT-cAMP sodium salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
8-CPT-cAMP sodium salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.