CAS RN 93840-60-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
1,3-Bis(2’bromo-4’-cyano-phenoxy)propane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.