missing translation for 'onlineSavingsMsg'
Få mere at vide

1,3-Bis(2’bromo-4’-cyano-phenoxy)propane, TRC

Artikelnummer. 30334458
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30334458 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30334458 Leverandør Toronto Research Chemicals Leverandørnr. TRCB41499010MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 1,3-Bis(2'bromo-4'-cyano-phenoxy)propane
CAS 93840-60-9
Molekylær formel C17 H12 Br2 N2 O2
Mængde 10 mg
Synonym 4,4'-[1,3-Propanediylbis(oxy)]bis[3-bromo-benzonitrile
InChI formel InChI=1S/C17H12Br2N2O2/c18-14-8-12(10-20)2-4-16(14)22-6-1-7-23-17-5-3-13(11-21)9-15(17)19/h2-5,8-9H,1,6-7H2
SMIL Brc1cc(ccc1OCCCOc2ccc(cc2Br)C#N)C#N
IUPAC navn 3-bromo-4-[3-(2-bromo-4-cyanophenoxy)propoxy]benzonitrile
Molekylvægt (g/mol) 436.097
Formel vægt 433.927
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.