CAS RN 936109-37-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
5
af
5
Resultater
1
Kampagner er tilgængelige
N-Methylperfluorooctanesulfonamide D3 (methyl D3) 50 μg/mL in Methanol:Water, Dr. Ehrenstorfer
N-Methylperfluorooctanesulfonamide D3 (methyl D3) 50 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
N-Methylperfluorooctanesulfonamide-d3, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
N-Methylperfluorooctanesulfonamide-d3, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5
N-Methylperfluorooctanesulfonamide-d3, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.