Få mere at vide
N-Methylperfluorooctanesulfonamide-d3 (50μg/mL in Methanol), TRC
Artikelnummer. 30472767
Skift visning
Mængde:
1 mL
Artikelnummer. Mængde
30472767 1 mL
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472767 Leverandør Toronto Research Chemicals Leverandørnr. TRCKIT79801X1ML
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methylperfluorooctanesulfonamide-d3
CAS 936109-37-4
Molekylær formel C9HD3F17NO2S
Mængde 1 mL
Synonym N-Methylperfluorooctane Sulfonamide D3 (methyl D3), 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluoro-N-(methyl-d3)-1-octanesulfonamide (ACI), 1-Octanesulfonamide, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-N-(methyl-d3)- (ACI), D3-N-MeFOSA, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluoro-N-(methyl-d3)-1-octanesulfonamide
InChI formel InChI=1S/C9H4F17NO2S/c1-27-30(28,29)9(25,26)7(20,21)5(16,17)3(12,13)2(10,11)4(14,15)6(18,19)8(22,23)24/h27H,1H3/i1D3
SMIL [2H]C([2H])([2H])NS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC navn 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-N-(trideuteriomethyl)octane-1-sulfonamide
Molekylvægt (g/mol) 516.19
Formel vægt 515.988
Koncentration eller sammensætning (efter analyt eller komponenter) N-Methylperfluorooctanesulfonamide-d3 50 μg/mL