CAS RN 934541-31-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
TAK-901, MedChemExpress
MedChemExpress TAK-901 is a multi-targeted aurora inhibitor with IC50s of 21 and 15 nM for aurora A and B, respectively.