CAS RN 930-27-8
IUPAC Navn:
3-methylfuran
Synonymer:
3-methylfuran
Molekylvægt (g/mol):
82.1
Molekylformel:
C5H6O
InChi Key:
KJRRQXYWFQKJIP-UHFFFAOYSA-N
SMILES:
CC1=COC=C1
1
–
5
af
5
Resultater
2
3-Methylfuran, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-Methylfuran, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
3-Methylfuran, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.