CAS RN 925-94-0
IUPAC Navn:
formic acid
Synonymer:
formic acid ameisensaeure hcooh acide formique bilorin H-cooh
Molekylvægt (g/mol):
46.03
Molekylformel:
CH2O2
InChi Key:
BDAGIHXWWSANSR-UHFFFAOYSA-N
SMILES:
OC=O
1
–
3
af
3
Resultater
1
Formic Acid-d, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Formic Acid-d, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Formic Acid-d, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.