CAS RN 92420-89-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
2,3,4-Tri-O-acetyl-a-D-glucuronic Acid Methyl Ester Trichloroacetimidate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,3,4-Tri-O-acetyl-a-D-glucuronic Acid Methyl Ester Trichloroacetimidate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,3,4-Tri-O-acetyl-a-D-glucuronic Acid Methyl Ester Trichloroacetimidate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.