CAS RN 92339-07-6
IUPAC Navn:
[2,3,5,6-tetrafluoro-4-(hydroxymethyl)phenyl]methanol
Synonymer:
[2,3,5,6-tetrafluoro-4-(hydroxymethyl)phenyl]methanol
Molekylvægt (g/mol):
210.13
Molekylformel:
C8H6F4O2
InChi Key:
SDHKGYDQOGCLQM-UHFFFAOYSA-N
SMILES:
OCC1=C(F)C(F)=C(CO)C(F)=C1F
1
–
1
af
1
Resultater
1
2,3,5,6-Tetrafluoro-1,4-benzenedimethanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality