CAS RN 92134-98-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Fosphenytoin disodium, MedChemExpress
MedChemExpress Fosphenytoin sodium is a phenytoin prodrug with similar anticonvulsant properties.