CAS RN 92-53-5
IUPAC Navn:
4-phenylmorpholine
Synonymer:
N-phenylmorpholine
Molekylvægt (g/mol):
163.22
Molekylformel:
C10H13NO
InChi Key:
FHQRDEDZJIFJAL-UHFFFAOYSA-N
SMILES:
C1CN(CCO1)C1=CC=CC=C1
1
–
3
af
3
Resultater
1
4-Phenylmorpholine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Phenylmorpholine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Phenylmorpholine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.