CAS RN 91221-46-4
IUPAC Navn:
4-[1-(4-hydroxyphenyl)-2-phenylbut-1-en-1-yl]phenol
Synonymer:
4-[1-(4-hydroxyphenyl)-2-phenylbut-1-en-1-yl]phenol
Molekylvægt (g/mol):
316.4
Molekylformel:
C22H20O2
InChi Key:
BPKSDMHGDYTXLI-UHFFFAOYSA-N
SMILES:
CCC(=C(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1)C1=CC=CC=C1
1
–
1
af
1
Resultater
1
1,1-Bis(4-hydroxyphenyl)-2-phenyl-1-butene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.