CAS RN 910297-59-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
2-[[6-[(2-Aminoethyl)amino]-2-methyl-4-pyrimidinyl]amino]-N-(2-chloro-6-methylphenyl)-5-thiazolecarboxamide, Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.