CAS RN 91-88-3
IUPAC Navn:
2-[ethyl(3-methylphenyl)amino]ethan-1-ol
Synonymer:
2-[ethyl(3-methylphenyl)amino]ethanol
Molekylvægt (g/mol):
179.26
Molekylformel:
C11H17NO
InChi Key:
KRNUKKZDGDAWBF-UHFFFAOYSA-N
SMILES:
CCN(CCO)C1=CC=CC(C)=C1
1
–
3
af
3
Resultater
1
2-(N-Ethyl-m-toluidino)ethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-(N-Ethyl-m-toluidino)ethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-(N-Ethyl-m-toluidino)ethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.