CAS RN 908337-63-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
2-Deoxy-2,2-difluoro-D-erythro-ribofuranose-3,5-dibenzoate 1-(2,2,2-Trichloroethanimidate), Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.