CAS RN 905858-63-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
N,N-Bis(methylsulphonyl)-2-phenoxyaniline, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
N-(Methylsulfonyl)-4-(desnitro) Nimesulide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-(Methylsulfonyl)-4-(desnitro) Nimesulide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
N-(Methylsulfonyl)-4-(desnitro) Nimesulide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.