Få mere at vide
N,N-Bis(methylsulphonyl)-2-phenoxyaniline, Mikromol™
Artikelnummer. 30051346
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051346 25 mg
30051042 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051346 Leverandør Mikromol Leverandørnr. MM0378.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N,N-Bis(methylsulphonyl)-2-phenoxyaniline
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N-Bis(methylsulphonyl)-2-phenoxyaniline
CAS 905858-63-1
Analyt- eller komponentnavne N,N-Bis(methylsulphonyl)-2-phenoxyaniline
Forsendelsestilstand Room Temperature
Molekylær formel C14 H15 N O5 S2
Certificeringer/compliance ISO 17025
Synonym N,N-Bis(methylsulphonyl)-2-phenoxyaniline, Nimesulide Imp. E (EP), Nimesulide Impurity E
InChI formel InChI=1S/C14H15NO5S2/c1-21(16,17)15(22(2,18)19)13-10-6-7-11-14(13)20-12-8-4-3-5-9-12/h3-11H,1-2H3
SMIL CS(N(S(C)(=O)=O)C1=C(OC2=CC=CC=C2)C=CC=C1)(=O)=O
IUPAC navn N-methylsulfonyl-N-(2-phenoxyphenyl)methanesulfonamide
Molekylvægt (g/mol) 341.4
Mængde 25 mg
Formel vægt 341.0392 g/mol