CAS RN 9052-95-3
IUPAC Navn:
1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene
Synonymer:
2-ethylstyrene; divinylbenzene; styrene
Molekylvægt (g/mol):
366.55
Molekylformel:
C28H30
InChi Key:
NWUYHJFMYQTDRP-UHFFFAOYSA-N
SMILES:
C=CC1=CC=CC=C1.CCC1=CC=CC=C1C=C.C=CC1=CC=CC=C1C=C
1
–
4
af
4
Resultater
1
Dianion HP-20 Resin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Diaion HP-20 Resin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Dianion HP-20 Resin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.