CAS RN 898-84-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
11beta-Hydroxyboldione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
11b-Hydroxyboldione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
11beta-Hydroxyandrosta-1,4-dien-3,17-dione (11beta-Hydroxyboldione), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4
11b-Hydroxyboldione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.