CAS RN 89678-08-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
6-Hydroxy-5-iodo-7-oxabicyclo[2.2.1]heptane-2-carboxylic Acid Gamma-Lactone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
6-Hydroxy-5-iodo-7-oxabicyclo[2.2.1]heptane-2-carboxylic Acid gamma-Lactone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
6-Hydroxy-5-iodo-7-oxabicyclo[2.2.1]heptane-2-carboxylic Acid Gamma-Lactone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.