CAS RN 89663-73-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
17-(Cyclopropylmethyl)-4'',4'',5'',5''-tetramethyl-4'',5''dihydro-(7betaH)-6alpha,14-ethano-(5betaH)-difurano[2',3',4',5':4,12,13,5;2'',3'':6,7]-14alpha-morphinan-3-ol, Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.