CAS RN 894351-86-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
N-Methyl-1-(phenylmethyl)-3-(1,2,3,6-tetrahydro-1-methyl-4-pyridinyl)-1H-indole-5-ethanesulfonamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Methyl-1-(phenylmethyl)-3-(1,2,3,6-tetrahydro-1-methyl-4-pyridinyl)-1H-indole-5-ethanesulfonamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.