CAS RN 888723-91-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(2S,3S,4aR,6S,8aR)-2,3,4a,5,6,8a-Hexahydro-7-iodo-2,3-dimethoxy-2,3,6-trimethyl-1,4-benzodioxin-6-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2S,3S,4aR,6S,8aR)-2,3,4a,5,6,8a-Hexahydro-7-iodo-2,3-dimethoxy-2,3,6-trimethyl-1,4-benzodioxin-6-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.