CAS RN 885704-21-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
c-Fms-IN-3, MedChemExpress
MedChemExpress c-Fms-IN-3 is a novel c-Fms kinase inhibitor with a potential as anti-inflammatory agent and antirheumatic agent.