CAS RN 881179-02-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Lith-O-Asp, MedChemExpress
MedChemExpress Lith-O-Asp is a sialytransferase (ST) inhibitor, with IC50s of 12-37 μM.