CAS RN 859853-30-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
BMS-690514, MedChemExpress
MedChemExpress BMS-690514 is a potent and orally active inhibitor of EGFR and VEGFR; has IC50s of 5, 20 and 60 nM for EGFR, HER 2 and HER 4, respectively.