CAS RN 85604-00-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Zaltidine, MedChemExpress
MedChemExpress Zaltidine(CP-57361) is a H2-receptor antagonist, which has the antisecretory action.