CAS RN 85259-19-4
IUPAC Navn:
methyl 2-bromo-2-(2-chlorophenyl)acetate
Synonymer:
methyl 2-bromo-2-(2-chlorophenyl)acetate
Molekylvægt (g/mol):
263.52
Molekylformel:
C9H8BrClO2
InChi Key:
HMBUCZUZRQQJQD-UHFFFAOYNA-N
SMILES:
COC(=O)C(Br)C1=CC=CC=C1Cl
1
–
3
af
3
Resultater
1
Methyl a-Bromo-2-chlorophenylacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Methyl a-Bromo-2-chlorophenylacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Methyl a-Bromo-2-chlorophenylacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.