CAS RN 85193-88-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
p-Nitrophenyl 2-Acetamido-2-deoxy-4-O-(2’,3’,4’,6’-tetra-O-acetyl-Beta-D-galactopyranosyl)-3,6-di-O-acetyl-Beta-D-glucopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
p-Nitrophenyl 2-Acetamido-2-deoxy-4-O-(2’,3’,4’,6’-tetra-O-acetyl-Beta-D-galactopyranosyl)-3,6-di-O-acetyl-Beta-D-glucopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
p-Nitrophenyl 2-Acetamido-2-deoxy-4-O-(2’,3’,4’,6’-tetra-O-acetyl-Beta-D-galactopyranosyl)-3,6-di-O-acetyl-Beta-D-glucopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.