CAS RN 851636-83-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ Cysmethynil
ICMT inhibitor; arrests cell-cycle at G1 phase