CAS RN 85081-18-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
(+)-alpha-Dihydrotetrabenazine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(+)-Alpha-Dihydrotetrabenazine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(+)-Alpha-Dihydrotetrabenazine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
NBI-98782, MedChemExpress
MedChemExpress NBI-98782(alpha-dihydrotetrabenazine) is a vesicular monoamine transporter (VMAT2) inhibtior with an Ki value of 0.97 nM.