CAS RN 849630-65-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
2-[(1RS)-8-Ethyl-1-(1-methylethyl)-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid (1-Isopropyl Etodolac), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.