CAS RN 84687-45-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Astragaloside VI, MedChemExpress
MedChemExpress Astragaloside VI could activate EGFR/ERK signalling pathway to improve wound healing.