CAS RN 84687-42-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Astragaloside III, MedChemExpress
MedChemExpress Astragaloside III is a natural product isolated from Astragalus.