CAS RN 84371-57-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(5a,10a,17b)-5,10-Epoxy-17-hydroxy-17-(1-propyn-1-yl)-estr-9(11)-en-3-one Cyclic 1,2-Ethanediyl Acetal, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(5Alpha,10Alpha,17Beta)-5,10-Epoxy-17-hydroxy-17-(1-propyn-1-yl)-estr-9(11)-en-3-one Cyclic 1,2-Ethanediyl Acetal, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(5Alpha,10Alpha,17Beta)-5,10-Epoxy-17-hydroxy-17-(1-propyn-1-yl)-estr-9(11)-en-3-one Cyclic 1,2-Ethanediyl Acetal, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.