CAS RN 84249-14-9
IUPAC Navn:
4-bromopyridin-2-amine
Synonymer:
4-bromopyridin-2-amine
Molekylvægt (g/mol):
173.01
Molekylformel:
C5H5BrN2
InChi Key:
BAQKUNMKVAPWGU-UHFFFAOYSA-N
SMILES:
NC1=CC(Br)=CC=N1
1
–
2
af
2
Resultater
1
2-Amino-4-bromopyridine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Amino-4-bromopyridine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.