CAS RN 84236-26-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(2S)-2-(5-Bromo-6-methoxynaphthalen-2-yl)propanoic Acid (5-Bromonaproxen), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
(S)-5-Bromo Naproxen, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(S)-5-Bromo Naproxen, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.