CAS RN 84-17-3
IUPAC Navn:
4-[(2E,4E)-4-(4-hydroxyphenyl)hexa-2,4-dien-3-yl]phenol
Synonymer:
dienestrol dienestrol, (Z,E)- para-dien dinestrol gynefollin sexadien (Z,Z)-isomer dienestrol dienoestrolum dienestrol, (E,E)- retalon
Molekylvægt (g/mol):
266.34
Molekylformel:
C18H18O2
InChi Key:
NFDFQCUYFHCNBW-SCGPFSFSSA-N
SMILES:
C\C=C(\C(=C\C)\C1=CC=C(O)C=C1)/C1=CC=C(O)C=C1
1
–
4
af
4
Resultater
1
Dienestrol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Dienestrol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Dienestrol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Dienestrol, MedChemExpress
MedChemExpress Dienestrol is a synthetic, non-steroidal estrogen, is an estrogen receptor agonist, for the treatment of menopausal and postmenopausal symptoms.