CAS RN 83493-63-4
IUPAC Navn:
2-(4-ethoxyphenyl)-2-methylpropan-1-ol
Synonymer:
2-(4-ethoxyphenyl)-2-methylpropan-1-ol
Molekylvægt (g/mol):
194.27
Molekylformel:
C12H18O2
InChi Key:
OZEZBKUHAGFQME-UHFFFAOYSA-N
SMILES:
CCOC1=CC=C(C=C1)C(C)(C)CO
1
–
3
af
3
Resultater
1
2-(4-Ethoxyphenyl)-2-methylpropanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-(4-Ethoxyphenyl)-2-methylpropanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-(4-Ethoxyphenyl)-2-methylpropanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.