CAS RN 83-63-6
IUPAC Navn:
N-acetyl-N-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}acetamide
Synonymer:
N-acetyl-N-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}acetamide
Molekylvægt (g/mol):
309.37
Molekylformel:
C18H19N3O2
InChi Key:
YIEDSISPYKQADU-UHFFFAOYSA-N
SMILES:
CC(=O)N(C(C)=O)C1=CC=C(C=C1C)N=NC1=CC=CC=C1C
1
–
1
af
1
Resultater
1
Diacetazotol, MedChemExpress
MedChemExpress Diacetazotol inhibits dioxin-induced ethoxyresorufin-O-deethylase (EROD) activity with IC50 of 75±4 nM. Diacetazotol extracts from patent US20070032458, compound 3.