CAS RN 82989-25-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tazanolast, MedChemExpress
MedChemExpress Tazanolast is a selective mast-cell-stabilizing drug, on ozone-induced airway hyperresponsiveness in guinea pigs.