CAS RN 826-39-1
IUPAC Navn:
hydrogen N,2,3,3-tetramethylbicyclo[2.2.1]heptan-2-amine chloride
Synonymer:
hydrogen mecamylamine chloride
Molekylvægt (g/mol):
203.75
Molekylformel:
C11H22ClN
InChi Key:
PKVZBNCYEICAQP-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].CNC1(C)C2CCC(C2)C1(C)C
1
–
4
af
4
Resultater
2
Mecamylamine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Mecamylamine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Mecamylamine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.