CAS RN 82549-51-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(6R,7R)-7-Amino-3-[(5-methyl-2H-tetrazol-2-yl)methyl]-8-oxo-5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(6R,7R)-7-Amino-3-[(5-methyl-2H-tetrazol-2-yl)methyl]-8-oxo-5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(6R,7R)-7-Amino-3-[(5-methyl-2H-tetrazol-2-yl)methyl]-8-oxo-5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.