CAS RN 82266-85-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Impurity C of Alfacalcidol, MedChemExpress
MedChemExpress Impurity of Alfacalcidol. Alfacalcidol (1-hydroxycholecalciferol; Alpha D3; 1.alpha.-Hydroxyvitamin D3) is a non-selective VDR activator medication.