CAS RN 819-79-4
IUPAC Navn:
bis(propan-2-yl)azanium chloride
Synonymer:
diisopropylazanium chloride
Molekylvægt (g/mol):
137.65
Molekylformel:
C6H16ClN
InChi Key:
URAZVWXGWMBUGJ-UHFFFAOYSA-N
SMILES:
[Cl-].CC(C)[NH2+]C(C)C
1
–
2
af
2
Resultater
1
Diisopropylamine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Diisopropylamine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.