CAS RN 81189-92-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Acetyllovastatin, MedChemExpress
MedChemExpress Acetyllovastatin, a acetate of Lovastatin, presentes a moderate inhibitory effect against the enzyme acetylcholinesterase with an IC50 of 79 μg/mL. Lovastatin has been found to display antifungal activity, and suppresses proliferation of a number of transformed cell lines.