CAS RN 81-19-6
IUPAC Navn:
1,3-dichloro-2-(dichloromethyl)benzene
Synonymer:
1,3-dichloro-2-(dichloromethyl)benzene
Molekylvægt (g/mol):
229.91
Molekylformel:
C7H4Cl4
InChi Key:
QQPXXHAEIGVZKQ-UHFFFAOYSA-N
SMILES:
ClC(Cl)C1=C(Cl)C=CC=C1Cl
1
–
2
af
2
Resultater
1
Alpha,Alpha,2,6-Tetrachloro-toluene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
α,α,2,6-Tetrachloro-toluene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.