Få mere at vide
Alpha,Alpha,2,6-Tetrachloro-toluene, TRC
Artikelnummer. 30414276
Skift visning
Mængde:
1 g
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30414276 2.5 g
30414158 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30414276 Leverandør Toronto Research Chemicals Leverandørnr. TRCT2920652.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale alpha,alpha-2,6-Tetrachlorotoluene
CAS 81-19-6
Molekylær formel C7 H4 Cl4
Mængde 2.5 g
Synonym alpha,alpha,2,6-Tetrachlorotoluene, Benzene','1,3-dichloro-2-(dichloromethyl)- (9CI','ACI), 1,3-Dichloro-2-(dichloromethyl)benzene (ACI), Toluene','α,α,2,6-tetrachloro- (7CI','8CI), 2,6,α,α-Tetrachlorotoluene, 2,6-Dichlorobenzal chloride, 2,6-Dichlorobenzylidene chloride, NSC 79866, α,α,2,6-Tetrachlorotoluene
InChI formel InChI=1S/C7H4Cl4/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3,7H
SMIL ClC(Cl)c1c(Cl)cccc1Cl
IUPAC navn 1,3-dichloro-2-(dichloromethyl)benzene
Molekylvægt (g/mol) 229.92
Formel vægt 227.9067